Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGK-beta Antibody (OTI1F9), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270552FR
Description
DGK-beta Monoclonal specifically detects DGK-beta in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot.Specifications
| DGK-beta | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| DGKB | |
| Full length recombinant protein of human DGK-beta (NP_663733) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Primate | |
| IgG1 |
| Western Blot | |
| OTI1F9 | |
| Western Blot | |
| 90 kDa diacylglycerol kinase, DAG kinase beta, DAGK2, DGK-beta, diacylglycerol kinase beta, diacylglycerol kinase, beta (90kD), diacylglycerol kinase, beta 90kDa, Diglyceride kinase beta, EC 2.7.1, EC 2.7.1.107, KIAA0718DGKDGK-BETA | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 1607.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction