Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGK-iota Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306558V
This item is not returnable.
View return policy
Description
DGK-iota Polyclonal antibody specifically detects DGK-iota in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
DGK-iota | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
DAG kinase iota, DGK-IOTA, diacylglycerol kinase iota, diacylglycerol kinase, iota, Diglyceride kinase iota, EC 2.7.1.107 | |
Produced in rabbits immunized with E. coli-derived Human DGK-iota fragment. | |
0.1 mL | |
Signal Transduction | |
9162 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction