Learn More
Invitrogen™ DGKA Polyclonal Antibody

Description
Immunogen sequence: KVSDVLKLFED GEMAKYVQGD AIGYEGFQQF LKIYLEVDNV PRHLSLALFQ SFETGHCLNE TNVTKDVVCL NDVSCYFSL Highest antigen sequence identity to the following orthologs - mouse 58%, rat 58%.
Specifications
Specifications
| Antigen | DGKA |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Dgka |
| Gene Accession No. | P23743 |
| Gene Alias | 80 kDa diacylglycerol kinase; 80kDa; AW146112; DAG kinase alpha; Dagk; Dagk1; Dgka; DGK-alpha; diacylglycerol kinase; diacylglycerol kinase alpha; diacylglycerol kinase alpha Flt2-Pr2; diacylglycerol kinase, alpha; diacylglycerol kinase, alpha 80kDa; Diglyceride kinase; diglyceride kinase alpha; MGC12821; MGC42356; truncated diacylglycerol kinase alpha Flt1-Pr1; truncated diacylglycerol kinase alpha Flt1-Pr2 |
| Gene Symbols | Dgka |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.