Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DGKB Monoclonal Antibody (OTI1F9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525167
Description
DGKB Monoclonal Antibody for Western Blot
Diacylglycerol (DAG) is an allosteric activator of protein kinase C. DAG also participates in regulating RAS and RHO family proteins by activating the guanine nucleotide exchange factors VAV and RASGRP1. DAG is also involved in the synthesis of phospholipids and triacylglycerols. Tight regulation of DAG levels is achieved via DAG kinases (DGKs), which remove DAG by phosphorylate it to phosphatidic acid. Several mammalian isozymes of DAGK have been identified.
Specifications
DGKB | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y6T7 | |
DGKB | |
Full length human recombinant protein of DGKB produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Canine, Monkey | |
Antibody | |
IgG1 |
Western Blot | |
OTI1F9 | |
Unconjugated | |
DGKB | |
6430574F24; 90 kDa diacylglycerol kinase; 90kda; C630029D13Rik; DAG kinase beta; Dagk; DAGK2; DGK; Dgkb; DGK-beta; Diacylglycerol kinase 90kDa; diacylglycerol kinase beta; diacylglycerol kinase beta, 90kDa; diacylglycerol kinase, beta; diacylglycerol kinase, beta 90kDa; diglyceride kinase beta; Kiaa0718; mKIAA0718 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1607, 482328 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction