Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGKB Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983238
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3238 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3238 Supplier Bioss Supplier No. BS14295R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Diacylglycerol (DAG) is an allosteric activator of protein kinase C. DAG also participates in regulating RAS and RHO family proteins by activating the guanine nucleotide exchange factors VAV and RASGRP1. DAG is also involved in the synthesis of phospholipids and triacylglycerols. Tight regulation of DAG levels is achieved via DAG kinases (DGKs), which remove DAG by phosphorylate it to phosphatidic acid. Several mammalian isozymes of DAGK have been identified.

Specifications

Antigen DGKB
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DGKB
Gene Accession No. Q9Y6T7
Gene Alias 6430574F24; 90 kDa diacylglycerol kinase; 90kda; C630029D13Rik; DAG kinase beta; Dagk; DAGK2; DGK; DGKB; DGK-beta; Diacylglycerol kinase 90kDa; Diacylglycerol kinase beta; diacylglycerol kinase beta, 90kDa; diacylglycerol kinase, beta; diacylglycerol kinase, beta 90kDa; Diglyceride kinase beta; KIAA0718; mKIAA0718
Gene Symbols DGKB
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DGKB.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1607
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.