Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DGKH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104522
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104522 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104522 Supplier Invitrogen™ Supplier No. PA5104522
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DGKH.

This gene encodes a member of the diacylglycerol kinase (DGK) enzyme family. Members of this family are involved in regulating intracellular concentrations of diacylglycerol and phosphatidic acid. Variation in this gene has been associated with bipolar disorder. Alternatively spliced transcript variants have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DGKH
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Dgkh
Gene Accession No. Q86XP1
Gene Alias 5930402B05Rik; D130015C16; DAG kinase eta; DGK; DGKeta; DGK-eta; DGKH; Diacylglycerol kinase eta; diacylglycerol kinase, eta; diglyceride kinase eta; DKFZp761I1510; EC 2.7.1.107; RGD1561955; si:ch211-93a19.1
Gene Symbols Dgkh
Host Species Rabbit
Immunogen A synthesized peptide derived from human DGKH(Accession Q86XP1), corresponding to amino acid residues V762-G812.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160851, 361076, 380921
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.