Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGKK Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983241
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3241 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3241 Supplier Bioss Supplier No. BS14300R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DGKK phosphorylates diacylglycerol (DAG) to generate phosphatidic acid (PA).

Specifications

Antigen DGKK
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DGKK
Gene Accession No. Q5KSL6
Gene Alias 142 kDa diacylglycerol kinase; DAG kinase kappa; DGKK; DGK-kappa; Diacylglycerol kinase kappa; diacylglycerol kinase, kappa; Diglyceride kinase kappa
Gene Symbols DGKK
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DGKK.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139189
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.