Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGKK Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | DGKK |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2691
![]() ![]() |
Proteintech Group Inc
200901AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to DGKK.
DGKK phosphorylates diacylglycerol (DAG) to generate phosphatidic acid (PA).Specifications
DGKK | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q5KSL6 | |
139189 | |
Peptide | |
Primary | |
-20°C | |
DGKK |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
142 kDa diacylglycerol kinase, DAG kinase kappa, DGK kappa, DGKK, Diacylglycerol kinase kappa, diacylglycerol kinase, kappa, Diglyceride kinase kappa | |
DGKK | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title