Learn More
DGKZ Mouse anti-Human, Clone: OTI1A6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809946

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified.Specifications
DGKZ | |
Monoclonal | |
Unconjugated | |
DGKZ | |
DAGK5, DAGK6, DGK-ZETA, hDGKzeta | |
Mouse | |
Affinity Chromatography | |
RUO | |
8525 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1A6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q13574 | |
DGKZ | |
Human recombit protein fragment corresponding to amino acids 734-808 of human DGKZ produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.