Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGKZ Polyclonal Antibody
DGKZ Polyclonal Antibody
Supplier: Thermo Scientific PA550613
Description
DGKZ Polyclonal Antibody for IHC (P)

Specifications
DGKZ | |
Polyclonal | |
Unconjugated | |
DGKZ | |
104 kDa diacylglycerol kinase; DAG kinase DAGK6; DAG kinase zeta; DAGK5; DAGK6; DGK-IV; DGKZ; DGK-zeta; Diacylglycerol kinase zeta; diacylglycerol kinase, zeta 104kDa; Diglyceride kinase; diglyceride kinase zeta; E130307B02Rik; ENSMUSG00000075035; F730209L11Rik; hDGKzeta; mDGK[z] | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104418, 81821, 8525 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O08560, Q13574, Q80UP3 | |
DGKZ | |
Synthetic peptide corresponding to residues near the C terminal of human diacylglycerol kinase, zeta. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction