Learn More
Invitrogen™ DGKZ Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120452
Description
Positive test controls include: LO2, HeLa, 293T, Mouse brain. The target is usually found in the following locations: Cell membrane, Cytoplasm, Nucleus. Immunogen sequence: ATMVQKAKRR SAAPLHSDQQ PVPEQLRIQV SRVSMHDYEA LHYDKEQLKE ASVPLGTVVV PGDSDLELCR AHIERLQQEP DGAGAKSPTC QKLSPKWCFL DATTASRFYR IDRAQEHLNY VTEIAQDEIY ILDPELLGAS ARPDLPTPTS PLPTSPCSPT PRSLQGDAAP PQGEELIEAA KRNDFCKLQE LHRAGGDLMH RDEQSRTLLH HAVSTGSKDV VRYLLDHAPP EILDAVEENG ETCLHQAAAL GQRTICHYIV EAGASLMKTD QQGDTPRQRA EKAQDTELAA YLENRQHYQM IQREDQETAV.
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified.
Specifications
DGKZ | |
Polyclonal | |
Unconjugated | |
DGKZ | |
104 kDa diacylglycerol kinase; DAG kinase DAGK6; DAG kinase zeta; DAGK5; DAGK6; DGK-IV; DGKZ; DGK-zeta; Diacylglycerol kinase zeta; diacylglycerol kinase, zeta 104kDa; Diglyceride kinase; diglyceride kinase zeta; E130307B02Rik; ENSMUSG00000075035; F730209L11Rik; hDGKzeta; mDGK[z] | |
Rabbit | |
Affinity Chromatography | |
RUO | |
104418, 8525 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q13574, Q80UP3 | |
DGKZ | |
Recombinant fusion protein containing a sequence corresponding to amino acids 808-1117 of human DGKZ (NP_001099010.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.