Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DH5a-lysate Rabbit anti-Bacteria, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 105591IG150UL

Description
DH5a is the most frequently used E. coli strain for routine cloning applications. In addition to supporting blue/white screening recA1 and endA1 mutations in DH5a increase insert stability and improve the quality of plasmid DNA prepared from minipreps.Specifications
DH5a-lysate | |
Polyclonal | |
Unconjugated | |
DH5a lysate | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Western Blot | |
0.7 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Rabbit | |
150 μL | |
Primary | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction