Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHCR24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592446
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592446 Supplier Invitrogen™ Supplier No. PA592446
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEIRIIPAKK YVKLRFEPVR GLEAICAKFT HESQRQENHF VEGLLYSLDE AVIMTGVMTD EAEPSKLNSI GNYYKPWFFK HVENYLKTNR EGLEYIPLRH YYHRHTRSIF WELQDIIPFG NNPIFRYLFG WMVPPKISLL KLTQGETLRK LYEQHHVVQD MLVPMKCLQQ ALHTFQNDIH VYPIWLCPFI LPSQPGLVHP KGNEAELYID IGAYGEPRVK HFEARSCMRQ LEKFVRSVHG FQMLYADCYM NREEFWEMFD GSLYHKLREK LGCQDAFPEV YDKICKAARH; Positive Samples: U-87MG, HT-29, PC-12, NCI-H460; Cellular Location: Endoplasmic reticulum membrane, Golgi apparatus membrane, Single-pass membrane protein.

DHCR24 is a flavin adenine dinucleotide (FAD)-dependent oxidoreductase which catalyzes the reduction of the delta-24 double bond of sterol intermediates during cholesterol biosynthesis. This protein contains a leader sequence that directs it to the endoplasmic reticulum membrane. Missense mutations in this gene have been associated with desmosterolosis. Also, reduced expression of its gene occurs in the temporal cortex of Alzheimer disease patients and overexpression has been observed in adrenal gland cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHCR24
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Dhcr24
Gene Accession No. Q15392, Q5BQE6, Q8VCH6
Gene Alias 2310076D10Rik; 24-dehydrocholesterol reductase; 3 beta-hydroxysterol delta 24-reductase; 3-beta-hydroxysterol delta-24 reductase; 3-beta-hydroxysterol delta-24-reductase; 5830417J06Rik; DCE; Delta(24)-sterol reductase; desmosterol-to-cholesterol enzyme; DHCR24; Diminuto/dwarf1 homolog; KIAA0018; mKIAA0018; Nbla03646; seladin 1; SELADIN1; seladin-1; selective AD indicator 1
Gene Symbols Dhcr24
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 227-516 of human DHCR24 (NP_0555771).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1718, 298298, 74754
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.