Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHCR7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550397
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550397 Supplier Invitrogen™ Supplier No. PA550397
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total DHCR7 protein.

Production of cholesterol by reduction of C7-C8 double bond of 7-dehydrocholesterol (7-DHC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHCR7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene DHCR7
Gene Accession No. O88455, Q9UBM7
Gene Alias 7-dehydrocholesterol reductase; 7-DHC reductase; AA409147; D7SR; delta-7-dehydrocholesterol reductase; Delta7-sterol reductase; Dhcr7; putative sterol reductase SR-2; SLOS; Sterol Delta(7)-reductase; sterol delta-7-reductase; Sterol reductase SR-2
Gene Symbols DHCR7
Host Species Rabbit
Immunogen Fusion protein corresponding to a region derived from internal residues of human 7-dehydrocholesterol reductase.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13360, 1717
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.