Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHCR7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577942 Supplier Invitrogen™ Supplier No. PA577942
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: HepG2 (boiled), HepG2 MF (boiled), HepG2 (unboiled), HepG2 MF (unboiled) Predicted Reactivity: Bovine (96%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Production of cholesterol by reduction of C7-C8 double bond of 7-dehydrocholesterol (7-DHC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHCR7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene DHCR7
Gene Accession No. O88455, Q9UBM7, Q9Z2Z8
Gene Alias 7-dehydrocholesterol reductase; 7-DHC reductase; AA409147; D7SR; delta-7-dehydrocholesterol reductase; Delta7-sterol reductase; Dhcr7; putative sterol reductase SR-2; SLOS; Sterol Delta(7)-reductase; sterol delta-7-reductase; Sterol reductase SR-2
Gene Symbols DHCR7
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human DHCR7. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13360, 1717, 64191
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.