Learn More
DHCR7 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305321AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Production of cholesterol by reduction of C7-C8 double bond of 7-dehydrocholesterol (7-DHC).Specifications
DHCR7 | |
Polyclonal | |
Unconjugated | |
DHCR7 | |
7-dehydrocholesterol reductase, 7-DHC reductase, Putative sterol reductase SR-2, Sterol Delta(7)-reductase, DHCR7, D7SR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1717 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9UBM7 | |
DHCR7 | |
Between 1 and 50 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.