Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHDDS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112400
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112400 Supplier Invitrogen™ Supplier No. PA5112400
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DHDDS Polyclonal Antibody for Western Blot, IHC (P), ELISA

Dehydrodolichyl diphosphate (dedol-PP) synthase catalyzes cis-prenyl chain elongation to produce the polyprenyl backbone of dolichol, a glycosyl carrier lipid required for the biosynthesis of several classes of glycoproteins. Dehydrodolichyl diphosphate (dedol-PP) synthase catalyzes cis-prenyl chain elongation to produce the polyprenyl backbone of dolichol, a glycosyl carrier lipid required for the biosynthesis of several classes of glycoproteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHDDS
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DHDDS
Gene Accession No. Q86SQ9
Gene Alias 3222401G21Rik; cis-IPTase; cis-isoprenyltransferase; cis-prenyl transferase; Cis-prenyltransferase subunit hCIT; CIT; CPT; dedol-PP synthase; dehydrodolichyl diphosphate syntase complex subunit Dhdds; dehydrodolichyl diphosphate synthase; dehydrodolichyl diphosphate synthase complex subunit DHDDS; dehydrodolichyl diphosphate synthase isoform a; dehydrodolichyl diphosphate synthase subunit; Dhdds; DS; epididymis tissue protein Li 189m; FLJ13102; HDS; RP59; W91638; wu:fd56c06; zgc:77088
Gene Symbols DHDDS
Host Species Rabbit
Immunogen Recombinant Human Dehydrodolichyl diphosphate synthase complex subunit DHDDS protein (1-333AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79947
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.