Learn More
Invitrogen™ DHDH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109632
Description
Immunogen sequence: MALRWGIVSV GLISSDFTAV LQTLPRSEHQ VVAVAARDLS RAKEFAQKHD IPKAYGSYEE LAKDPSVEVA YIGTQHPQHK AAVMLCLAAG KAVLCEKPTG VNAAEVREMV AEARSRALFL MEAIWTRFFP ASEALRSVLA QGTLGDLRVA RAEFGKNLIH VPRAVDRAQA GGALLDIGIY CVQFTSMVFG GQKPEKISVV GRRHETGVDD TVTVLLQYPG EVHGSFTCSI TVQLSNTASV SGTKGMVQLL NPCWCPTELV VKGEHKEFPL PPVPKDCNFD NGAGMSYEAK HVWECLRKGM KESPVIPLSE SELLADILEE VRKAIGVTFP QDKR.
This gene encodes an enzyme that belongs to the family of dihydrodiol dehydrogenases, which exist in multiple forms in mammalian tissues and are involved in the metabolism of xenobiotics and sugars. These enzymes catalyze the NADP1-linked oxidation of transdihydrodiols of aromatic hydrocarbons to corresponding catechols. This enzyme is a dimeric dihydrodiol dehydrogenase, and it differs from monomeric dihydrodiol dehydrogenases in its high substrate specificity for trans-dihydrodiols of aromatic hydrocarbons in the oxidative direction.
Specifications
| DHDH | |
| Polyclonal | |
| Unconjugated | |
| DHDH | |
| 1300018L09Rik; 2DD; 3-deoxyglucosone reductase; AA591799; CAN2DD; ch211-203b17.3; DDH; DHDH; dhdh.2; dhdh-like.1.L; dihydrodiol dehydrogenase; dihydrodiol dehydrogenase (dimeric); dihydrodiol dehydrogenase, tandem duplicate 2; dimeric dihydrodiol dehydrogenase; D-xylose 1-dehydrogenase; D-xylose-NADP dehydrogenase; Hum2DD; putative LOC617564 protein; SUS2DD; Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase L homeolog; XELAEV_18036112mg; Xld1; Xld-1; xylose dehydrogenase 1; zgc:101723 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 71755 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.94 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9DBB8 | |
| DHDH | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-334 of human DHDH (NP_055290.1). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.