Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHDH Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS14302R

Description
This gene encodes an enzyme that belongs to the family of dihydrodiol dehydrogenases, which exist in multiple forms in mammalian tissues and are involved in the metabolism of xenobiotics and sugars. These enzymes catalyze the NADP1-linked oxidation of transdihydrodiols of aromatic hydrocarbons to corresponding catechols. This enzyme is a dimeric dihydrodiol dehydrogenase, and it differs from monomeric dihydrodiol dehydrogenases in its high substrate specificity for trans-dihydrodiols of aromatic hydrocarbons in the oxidative direction.Specifications
DHDH | |
Polyclonal | |
Unconjugated | |
DHDH | |
1300018L09Rik; 2DD; 3-deoxyglucosone reductase; AA591799; DDH; Dhdh; dihydrodiol dehydrogenase; dihydrodiol dehydrogenase (dimeric); Dimeric dihydrodiol dehydrogenase; D-xylose 1-dehydrogenase; D-xylose-NADP dehydrogenase; Hum2DD; Sus2DD; Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; Xld1; Xld-1; xylose dehydrogenase 1 | |
Rabbit | |
Protein A | |
RUO | |
27294 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9UQ10 | |
DHDH | |
KLH conjugated synthetic peptide derived from human DHDH. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction