Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHFR Monoclonal Antibody (OTI3A11)
GREENER_CHOICE

Catalog No. PIMA524945
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524945 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524945 Supplier Invitrogen™ Supplier No. MA524945
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DHFR Monoclonal Antibody for Western Blot, IHC (P)

Dihydrofolate reductase converts dihydrofolate into tetrahydrofolate, and is required for the synthesis of purines, thymidylic acid and certain amino acids. While the functional dihydrofolate reductase gene has been mapped to chromosome 5, multiple intronless processed pseudogenes or dihydrofolate reductase like genes have been identified on separate chromosomes. Dihydrofolate reductase deficiency has been linked to megaloblastic anemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHFR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DHFR
Gene Accession No. P00374
Gene Alias 8430436I03Rik; AA607882; AI662710; AW555094; Dhfr; Dhfr1; DHFRP1; Dihydrofolate reductase; Dihydrofolate reductase 1 (active); DYR
Gene Symbols DHFR
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human DHFR expression vector.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1719
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.