Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHFR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579152
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579152 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579152 Supplier Invitrogen™ Supplier No. PA579152
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human Jurkat whole cell, rat liver tissue, rat kidney tissue, mouse liver tissue, mouse kidney tissue. IHC: human colorectal cancer tissue, mouse small intestine tissue, rat small intestine tissue. ICC/IF: U20S cell. Flow: PC-3 cell.

Dihydrofolate reductase converts dihydrofolate into tetrahydrofolate, and is required for the synthesis of purines, thymidylic acid and certain amino acids. While the functional dihydrofolate reductase gene has been mapped to chromosome 5, multiple intronless processed pseudogenes or dihydrofolate reductase like genes have been identified on separate chromosomes. Dihydrofolate reductase deficiency has been linked to megaloblastic anemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHFR
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.01mg sodium azide
Gene DHFR
Gene Accession No. P00374, P00375, Q920D2
Gene Alias 8430436I03Rik; AA607882; AI662710; AW555094; Dhfr; Dhfr1; DHFRP1; Dihydrofolate reductase; Dihydrofolate reductase 1 (active); DYR
Gene Symbols DHFR
Host Species Rabbit
Immunogen E.coli-derived human DHFR recombinant protein (Position: V2-D187).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13361, 1719, 24312
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.