Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DHFR Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115117
Description
Antibody detects endogenous levels of total DHFR.
Dihydrofolate reductase converts dihydrofolate into tetrahydrofolate, and is required for the synthesis of purines, thymidylic acid and certain amino acids. While the functional dihydrofolate reductase gene has been mapped to chromosome 5, multiple intronless processed pseudogenes or dihydrofolate reductase like genes have been identified on separate chromosomes. Dihydrofolate reductase deficiency has been linked to megaloblastic anemia.
Specifications
| DHFR | |
| Polyclonal | |
| Unconjugated | |
| DHFR | |
| 8430436I03Rik; AA607882; AI662710; AW555094; Dhfr; Dhfr1; DHFRP1; Dihydrofolate reductase; Dihydrofolate reductase 1 (active); DYR | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13361, 1719, 24312 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P00374, P00375, Q920D2 | |
| DHFR | |
| A synthesized peptide derived from human DHFR(Accession P00374), corresponding to amino acid residues C7-T57. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction