Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHFRL1 Antibody (OTI1E2), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272459MFV610
Description
DHFRL1 Monoclonal antibody specifically detects DHFRL1 in Human, Mouse, Canine, Monkey samples. It is validated for Flow CytometrySpecifications
DHFRL1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
200895 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
OTI1E2 | |
Flow Cytometry | |
DHFRP4, dihydrofolate reductase pseudogene 4, dihydrofolate reductase-like 1, dihydrofolate reductase-like protein 1, EC 1.5.1.-, EC 1.5.1.3, FLJ16119 | |
Human recombinant protein fragment corresponding to amino acids 4-185 of human DHFRL1(NP_789785) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Canine, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction