Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHFRL1 Monoclonal Antibody (OTI1E2)
GREENER_CHOICE

Catalog No. PIMA525633
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525633 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525633 Supplier Invitrogen™ Supplier No. MA525633
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DHFRL1 Monoclonal Antibody for Western Blot, ICC/IF

Key enzyme in folate metabolism. Contributes to the de novo mitochondrial thymidylate biosynthesis pathway. Required to prevent uracil accumulation in mtDNA. Binds its own mRNA and that of DHFR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHFRL1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1E2
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DHFR2
Gene Accession No. Q86XF0
Gene Alias DHFR2; DHFRL1; DHFRP4; dihydrofolate reductase 2; Dihydrofolate reductase 2, mitochondrial; dihydrofolate reductase like 1; dihydrofolate reductase pseudogene 4; dihydrofolate reductase, mitochondrial; dihydrofolate reductase-like 1; dihydrofolate reductase-like protein 1; LOW QUALITY PROTEIN: dihydrofolate reductase 2, mitochondrial
Gene Symbols Dhfr-rs1, DHFR2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 4-185 of DHFRL1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 111208, 200895
Target Species Human, Mouse, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.