Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHFRL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548563
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548563 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548563 Supplier Invitrogen™ Supplier No. PA548563
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DHFRL1 Polyclonal Antibody for Western Blot, IHC (P)

Key enzyme in folate metabolism. Contributes to the de novo mitochondrial thymidylate biosynthesis pathway. Required to prevent uracil accumulation in mtDNA. Binds its own mRNA and that of DHFR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHFRL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DHFR2
Gene Accession No. Q86XF0
Gene Alias DHFR2; DHFRL1; DHFRP4; dihydrofolate reductase 2; Dihydrofolate reductase 2, mitochondrial; dihydrofolate reductase like 1; dihydrofolate reductase pseudogene 4; dihydrofolate reductase, mitochondrial; dihydrofolate reductase-like 1; dihydrofolate reductase-like protein 1; LOW QUALITY PROTEIN: dihydrofolate reductase 2, mitochondrial
Gene Symbols DHFR2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 45-74 amino acids from the Central region of human DHFRL1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200895
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.