Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DHH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5122037
This item is not returnable.
View return policy
Description
DHH Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a member of the Hedgehog family. The hedgehog gene family encodes signaling molecules that play an important role in regulating morphogenesis. This protein is predicted to be made as a precursor that is autocatalytically cleaved; the N-terminal portion is soluble and contains the signaling activity while the C-terminal portion is involved in precursor processing. More importantly, the C-terminal product covalently attaches a cholesterol moiety to the N-terminal product, restricting the N-terminal product to the cell surface and preventing it from freely diffusing throughout the organism. Defects in this protein have been associated with partial gonadal dysgenesis (PGD) accompanied by minifascicular polyneuropathy. This protein may be involved in both male gonadal differentiation and perineurial development.
Specifications
DHH | |
Polyclonal | |
Unconjugated | |
Dhh | |
C78960; desert hedgehog; desert hedgehog protein; Desert hedgehog protein C-product; Desert hedgehog protein N-product; Dhh; GDXYM; HHG-3; SRXY7 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
13363 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q61488 | |
Dhh | |
Recombinant protein Hedgehog Homolog, Desert. The antigen corresponds to amino acid range 212-392 of the target protein. | |
20 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction