Learn More
Invitrogen™ DHH Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547229
Description
In Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Dhh N-terminus peptide (aa 23-198), rmShh C-terminus peptide (aa 199-437), and rmIhh C-terminus peptide (aa 241-449) is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
This gene encodes a member of the Hedgehog family. The hedgehog gene family encodes signaling molecules that play an important role in regulating morphogenesis. This protein is predicted to be made as a precursor that is autocatalytically cleaved; the N-terminal portion is soluble and contains the signaling activity while the C-terminal portion is involved in precursor processing. More importantly, the C-terminal product covalently attaches a cholesterol moiety to the N-terminal product, restricting the N-terminal product to the cell surface and preventing it from freely diffusing throughout the organism. Defects in this protein have been associated with partial gonadal dysgenesis (PGD) accompanied by minifascicular polyneuropathy. This protein may be involved in both male gonadal differentiation and perineurial development.
Specifications
DHH | |
Polyclonal | |
Unconjugated | |
Dhh | |
C78960; desert hedgehog; desert hedgehog protein; Desert hedgehog protein C-product; Desert hedgehog protein N-product; Dhh; GDXYM; HHG-3; SRXY7 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
13363 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q61488 | |
Dhh | |
E. coli-derived recombinant mouse Desert Hedgehog/Dhh C-Terminus Cys199-Gly396. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.