Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109633
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109633 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109633 Supplier Invitrogen™ Supplier No. PA5109633
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CFPGNATVRL WSGERKGLRE LHRGDWVLAA DASGRVVPTP VLLFLDRDLQ RRASFVAVET EWPPRKLLLT PWHLVFAARG PAPAPGDFAP VFARRLRAGD SVLAPGGDAL RPARVARVAR EEAVGVFAPL TAHGTLLVND VLASCYAVLE SHQWAHRAFA PLRLLHALGA LLPGGAVQPT GMHWYSRLLY RLAEELLG.

This gene encodes a member of the Hedgehog family. The hedgehog gene family encodes signaling molecules that play an important role in regulating morphogenesis. This protein is predicted to be made as a precursor that is autocatalytically cleaved; the N-terminal portion is soluble and contains the signaling activity while the C-terminal portion is involved in precursor processing. More importantly, the C-terminal product covalently attaches a cholesterol moiety to the N-terminal product, restricting the N-terminal product to the cell surface and preventing it from freely diffusing throughout the organism. Defects in this protein have been associated with partial gonadal dysgenesis (PGD) accompanied by minifascicular polyneuropathy. This protein may be involved in both male gonadal differentiation and perineurial development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Dhh
Gene Accession No. O43323, Q61488
Gene Alias C78960; desert hedgehog; desert hedgehog protein; Desert hedgehog protein C-product; Desert hedgehog protein N-product; Dhh; GDXYM; HHG-3; SRXY7
Gene Symbols Dhh
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 199-396 of human DHH (NP_066382.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13363, 50846, 84380
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.