Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHODH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552694
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552694 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552694 Supplier Invitrogen™ Supplier No. PA552694
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QERDALRRVH RPAVLVKIAP DLTSQDKEDI ASVVKELGID GLIVTNTTVS RPAGLQGALR SETGGLSGKP LRDLSTQTIR EMYALTQGRV PIIGVGGVSS GQDALEKIRA G Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.

The protein encoded by this gene catalyzes the fourth enzymatic step, the ubiquinone-mediated oxidation of dihydroorotate to orotate, in de novo pyrimidine biosynthesis. This protein is a mitochondrial protein located on the outer surface of the inner mitochondrial membrane. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHODH
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DHODH
Gene Accession No. Q02127
Gene Alias 2810417D19Rik; AI834883; DHOdehase; DHODH; dihydroorotate dehydrogenase; dihydroorotate dehydrogenase (quinone); dihydroorotate dehydrogenase (quinone), mitochondrial; dihydroorotate dehydrogenase, mitochondrial; dihydroorotate oxidase; human complement of yeast URA1; POADS; URA1
Gene Symbols DHODH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DHODH. Recombinant protein control fragment (Product #RP-90071).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1723
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.