Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DHODH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552880
Description
Immunogen sequence: LAVRFTSLGL LPRARFQDSD MLEVRVLGHK FRNPVGIAAG FDKHGEAVDG LYKMGFGFVE IGSVTPKPQE GNPRPRVFRL PEDQAVINRY GFNSHGLSVV EHRLRG Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
The protein encoded by this gene catalyzes the fourth enzymatic step, the ubiquinone-mediated oxidation of dihydroorotate to orotate, in de novo pyrimidine biosynthesis. This protein is a mitochondrial protein located on the outer surface of the inner mitochondrial membrane. [provided by RefSeq, Jul 2008].
Specifications
| DHODH | |
| Polyclonal | |
| Unconjugated | |
| DHODH | |
| 2810417D19Rik; AI834883; DHOdehase; DHODH; dihydroorotate dehydrogenase; dihydroorotate dehydrogenase (quinone); dihydroorotate dehydrogenase (quinone), mitochondrial; dihydroorotate dehydrogenase, mitochondrial; dihydroorotate oxidase; human complement of yeast URA1; POADS; URA1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1723 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q02127 | |
| DHODH | |
| Recombinant protein corresponding to Human DHODH. Recombinant protein control fragment (Product #RP-90070). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction