Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHODH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589141
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589141 Supplier Invitrogen™ Supplier No. PA589141
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EIGSVTPKPQ EGNPRPRVFR LPEDQAVINR YGFNSHGLSV VEHRLRARQQ KQAKLTEDGL PLGVNLGKNK TSVDAAEDYA EGVRVLGPLA DYLVVNVSSP NTAGLRSLQG KAELRRLLTK VLQERDGLRR VHRPAVLVKI APDLTSQDKE DIASVVKELG IDGLIVTNTT VSRPAGLQGA LRSETGGLSG KPLRDLSTQT IREMYALTQG RVPIIGVGGV SSGQDALEKI RAGASLVQLY TALTFWGPPV VGKVKRELEA LLKEQGFGGV TDAIGADHRR; Positive Samples: HL-60, SW620, HepG2, Mouse heart, Mouse spleen, Mouse kidney, Rat heart, Rat kidney; Cellular Location: Mitochondrion inner membrane, Single-pass membrane protein.

The protein encoded by this gene catalyzes the fourth enzymatic step, the ubiquinone-mediated oxidation of dihydroorotate to orotate, in de novo pyrimidine biosynthesis. This protein is a mitochondrial protein located on the outer surface of the inner mitochondrial membrane. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHODH
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DHODH
Gene Accession No. O35435, Q02127, Q63707
Gene Alias 2810417D19Rik; AI834883; DHOdehase; DHODH; dihydroorotate dehydrogenase; dihydroorotate dehydrogenase (quinone); dihydroorotate dehydrogenase (quinone), mitochondrial; dihydroorotate dehydrogenase, mitochondrial; dihydroorotate oxidase; human complement of yeast URA1; POADS; URA1
Gene Symbols DHODH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 116-395 of human DHODH (NP_0013522).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1723, 56749, 65156
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.