Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHPS Antibody (OTI2C9), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270570APC
Description
DHPS Monoclonal antibody specifically detects DHPS in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
DHPS | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
OTI2C9 | |
Western Blot | |
deoxyhypusine synthase, DHS, DS, EC 2.5.1, EC 2.5.1.46, hypusine, MIG13, migration-inducing gene 13 | |
Full length human recombinant protein of human DHPS (NP_001921) produced in HEK293T cell. | |
0.1 mL | |
Cell Cycle and Replication | |
1725 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction