Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHPS Polyclonal Antibody
DHPS Polyclonal Antibody
Supplier: Thermo Scientific PA5101552
Description
DHPS Polyclonal Antibody for Western Blot, ICC/IF

Specifications
DHPS | |
Polyclonal | |
Unconjugated | |
Dhps | |
Deoxyhypusine synthase; DHPS; DHS; DS; MIG13; migration-inducing gene 13 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1725, 330817 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P49366, Q3TXU5 | |
Dhps | |
A synthesized peptide derived from human DHPS(Accession P49366), corresponding to amino acid residues F54-T104. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction