Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHRS1 Monoclonal Antibody (OTI4C12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA812489

Description
This gene encodes a member of the short-chain dehydrogenases/reductases (SDR) family. The encoded enzyme contains a conserved catalytic domain and likely functions as an oxidoreductase. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
DHRS1 | |
Monoclonal | |
Unconjugated | |
DHRS1 | |
SDR19C1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
115817 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI4C12 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96LJ7 | |
DHRS1 | |
Full length human recombit protein of human DHRS1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction