Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHRS11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561691
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561691 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561691 Supplier Invitrogen™ Supplier No. PA561691
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGLRQELREA QTHIRATCIS PGVVETQFAF KLHDKDPEKA AATYEQMKCL KPEDVAEAVI YVLSTPAHIQ IGDIQMRPTE QVT Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

DHRS11 gene ontology annotations related to this gene include oxidoreductase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DHRS11
Gene Accession No. Q6UWP2
Gene Alias 17-beta-hydroxysteroid dehydrogenase; 3-beta-hydroxysteroid 3-dehydrogenase; ARPG836; dehydrogenase/reductase (SDR family) member 11; dehydrogenase/reductase 11; dehydrogenase/reductase SDR family member 11; DHRS11; Estradiol 17-beta-dehydrogenase; hypothetical protein LOC510264; inactive dehydrogenase/reductase isoform; LOW QUALITY PROTEIN: dehydrogenase/reductase SDR family member 11; RGD1307935; SDR24C1; short chain dehydrogenase/reductase family 24C, member 1; short-chain dehydrogenase/reductase family 24C member 1; spDHRS11; UNQ836/PRO1774
Gene Symbols DHRS11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DHRS11. Recombinant protein control fragment (Product #RP-98379).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.