Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHRS11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589814
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589814 Supplier Invitrogen™ Supplier No. PA589814
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MARPGMERWR DRLALVTGAS GGIGAAVARA LVQQGLKVVG CARTVGNIEE LAAECKSAGY PGTLIPYRCD LSNEEDILSM FSAIRSQHSG VDICINNAGL ARPDTLLSGS TSGWKDMFNV NVLALSICTR EAYQSMKERN VDDGHIININ SMSGHRVLPL SVTHFYSATK YAVTALTEGL RQELREAQTH IRATCISPGV VETQFAFKLH DKDPEKAAAT YEQMKCLKPE DVAEAVIYVL STPAHIQIGD IQMRPTEQVT; Positive Samples: HT-29, HeLa, Mouse heart, Mouse kidney, Mouse lung, Mouse spleen.

DHRS11 gene ontology annotations related to this gene include oxidoreductase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS11
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DHRS11
Gene Accession No. Q3U0B3, Q6UWP2
Gene Alias 17-beta-hydroxysteroid dehydrogenase; 3-beta-hydroxysteroid 3-dehydrogenase; ARPG836; dehydrogenase/reductase (SDR family) member 11; dehydrogenase/reductase 11; dehydrogenase/reductase SDR family member 11; DHRS11; Estradiol 17-beta-dehydrogenase; hypothetical protein LOC510264; inactive dehydrogenase/reductase isoform; LOW QUALITY PROTEIN: dehydrogenase/reductase SDR family member 11; RGD1307935; SDR24C1; short chain dehydrogenase/reductase family 24C, member 1; short-chain dehydrogenase/reductase family 24C member 1; spDHRS11; UNQ836/PRO1774
Gene Symbols DHRS11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human DHRS11 (NP_0772842).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192970, 79154
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.