Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHRS12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566158
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566158 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566158 Supplier Invitrogen™ Supplier No. PA566158
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLHVKTLSLMT WRSRFLEESF WSLEETAALA KQLPLKSPSE NIFLHIVDLS DPKQIWKFVE NFKQEHKLHV LI Highest antigen sequence identity to the following orthologs - mouse 26%, rat 26%.

Putative oxidoreductase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS12
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DHRS12
Gene Accession No. A0PJE2
Gene Alias dehydrogenase/reductase (SDR family) member 12; dehydrogenase/reductase 12; dehydrogenase/reductase SDR family member 12; DHRS12; SDR40C1; short chain dehydrogenase/reductase family 40C, member 1; short-chain dehydrogenase/reductase family 40C member 1; zgc:153679
Gene Symbols DHRS12
Host Species Rabbit
Immunogen Recombinant Human DHRS12. Recombinant protein control fragment (Product #RP-106843).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79758
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.