Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHRS13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554657 Supplier Invitrogen™ Supplier No. PA554657
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAHRLWEASK RLAGLGPGED AEPDEDPQSE DSEAPSSLST PHPEEPTVSQ PYPSPQSSPD LSKMTHRIQA KVEPEIQLS Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 51%.

Putative oxidoreductase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS13
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DHRS13
Gene Accession No. Q6UX07
Gene Alias 2610209N15Rik; androgen-regulated short-chain dehydrogenase/reductase 1; prostate short-chain dehydrogenase reductase 1; likely ortholog of mouse cell line MC/9.IL4 derived transcript 1; CGI-82 protein; HCV core-binding protein; dehydrogenase/reductase (SDR family) member 13; dehydrogenase/reductase 13; dehydrogenase/reductase SDR family member 13; Dhrs13; RGD1305508; SDR7C5; Short chain dehydrogenase/reductase family 7C member 5; short chain dehydrogenase/reductase family 7C, member 5; UNQ419/PRO853
Gene Symbols DHRS13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DHRS13. Recombinant protein control fragment (Product #RP-93375).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147015
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.