Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHRS13 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988412
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8412 Supplier Bioss Supplier No. BS8261R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Putative oxidoreductase.

Specifications

Antigen DHRS13
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DHRS13
Gene Accession No. Q6UX07
Gene Alias dehydrogenase/reductase (SDR family) member 13; dehydrogenase/reductase 13; Dehydrogenase/reductase SDR family member 13; DHRS13; SDR7C5; Short chain dehydrogenase/reductase family 7C member 5; short chain dehydrogenase/reductase family 7C, member 5; UNQ419/PRO853
Gene Symbols DHRS13
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DHRS13.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147015
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.