Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHRS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564419
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564419 Supplier Invitrogen™ Supplier No. PA564419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDREQLVAKA LEHCGGVDFL VCSAGVNPLV GSTLGTSEQI WDKILSVNVK SPALLLSQL Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.

DHRS2 displays NADPH-dependent dicarbonyl reductase activity in vitro with 3,4-Hexanedione, 2,3-Heptanedione and 1-Phenyl-1,2-propanedione as substrates. DHRS2 do not reductase activity is displayed in vitro with steroids, retinoids and sugars as substrates. This protein may inhibit cell replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DHRS2
Gene Accession No. Q13268
Gene Alias 5430405K24Rik; dehydrogenase/reductase (SDR family) member 2; dehydrogenase/reductase 2; dehydrogenase/reductase member 2; dehydrogenase/reductase SDR family member 2, mitochondrial; Dhrs2; Dicarbonyl reductase HEP27; Hep27; Protein D; SDR family member; SDR25C1; Short chain dehydrogenase/reductase family 25C member 1; short chain dehydrogenase/reductase family 25C, member 1; short-chain alcohol dehydrogenase family member
Gene Symbols DHRS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DHRS2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10202
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.