Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHRS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552750 Supplier Invitrogen™ Supplier No. PA552750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TEKCLKETTE EIRQMGTECH YFICDVGNRE EVYQTAKAVR EKVGDITILV NNAAVVHGKS LMDSDDDALL KSQHINTLGQ FWTTKAFLPR MLELQNGHIV CLNSVLALSA IPG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

DHRS3 catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dhrs3
Gene Accession No. O75911, O88876
Gene Alias DD83.1; dehydrogenase/reductase (SDR family) member 3; dehydrogenase/reductase 3; dehydrogenase/reductase member 3; Dhrs3; RDH17; retinal dehydrogenase/reductase family member 3; retinal short-chain dehydrogenase/reductase 1; Retinol dehydrogenase 17; retSDR1; Rsdr1; SDR1; SDR16C1; Short chain dehydrogenase/reductase family 16C member 1; short chain dehydrogenase/reductase family 16C, member 1; short-chain dehydrogenase/reductase 1; short-chain dehydrogenase/reductase 3; UNQ2424/PRO4983
Gene Symbols Dhrs3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DHRS3. Recombinant protein control fragment (Product #RP-90377).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20148, 313689, 9249
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.