Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHRS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106292 Supplier Thermo Scientific Supplier No. PA5106292
Add to Cart
Add to Cart

DHRS3 Polyclonal Antibody

DHRS3 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Dhrs3
Gene Accession No. O75911, O88876
Gene Alias DD83.1; dehydrogenase/reductase (SDR family) member 3; dehydrogenase/reductase 3; dehydrogenase/reductase member 3; Dhrs3; RDH17; retinal dehydrogenase/reductase family member 3; retinal short-chain dehydrogenase/reductase 1; Retinol dehydrogenase 17; retSDR1; Rsdr1; SDR1; SDR16C1; Short chain dehydrogenase/reductase family 16C member 1; short chain dehydrogenase/reductase family 16C, member 1; short-chain dehydrogenase/reductase 1; short-chain dehydrogenase/reductase 3; UNQ2424/PRO4983
Gene Symbols Dhrs3
Host Species Rabbit
Immunogen A synthesized peptide derived from human DHRS3(Accession O75911), corresponding to amino acid residues A59-A109.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20148, 313689, 9249
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.