Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DHRS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112401
Description
DHRS3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
DHRS3 catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids.
Specifications
DHRS3 | |
Polyclonal | |
Unconjugated | |
Dhrs3 | |
DD83.1; dehydrogenase/reductase (SDR family) member 3; dehydrogenase/reductase 3; dehydrogenase/reductase member 3; Dhrs3; RDH17; retinal dehydrogenase/reductase family member 3; retinal short-chain dehydrogenase/reductase 1; Retinol dehydrogenase 17; retSDR1; Rsdr1; SDR1; SDR16C1; Short chain dehydrogenase/reductase family 16C member 1; short chain dehydrogenase/reductase family 16C, member 1; short-chain dehydrogenase/reductase 1; short-chain dehydrogenase/reductase 3; UNQ2424/PRO4983 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20148, 9249 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O75911, O88876 | |
Dhrs3 | |
Recombinant Human Short-chain dehydrogenase/reductase 3 protein (30-169AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction