Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHRS4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587775
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587775 Supplier Invitrogen™ Supplier No. PA587775
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MHKAGLLGLC ARAWNSVRMA SSGMTRRDPL ANKVALVTAS TDGIGFAIAR RLAQDGAHVV VSSRKQQNVD QAVATLQGEG LSVTGTVCHV GKAEDRERLV ATAVKLHGGI DILVSNAAVN PFFGSIMDVT EEVWDKTLDI NVKAPALMTK AVVPEMEKRG GGSVVIVSSI AAFSPSPGFS PYNVSKTALL GLTKTLAIEL APRNIRVNCL APGLIKTSFS RMLWMDKEKE ESMKETLRIR RLGEPEDCAG IVSFLCSEDA SYITGETVVV GGGTPSRL; Positive Samples: HeLa, LO2, A-549, MCF-7, SW620, Mouse liver, Mouse lung, Mouse intestine, Rat liver; Cellular Location: Nucleus, Peroxisome.

Reduces all-trans-retinal and 9-cis retinal. Can also catalyze the oxidation of all-trans-retinol with NADP as co-factor, but with much lower efficiency. Reduces alkyl phenyl ketones and alpha-dicarbonyl compounds with aromatic rings, such as pyrimidine-4-aldehyde, 3-benzoylpyridine, 4-benzoylpyridine, menadione and 4-hexanoylpyridine. Has no activity towards aliphatic aldehydes and ketones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DHRS4
Gene Accession No. Q8VID1, Q99LB2, Q9BTZ2
Gene Alias AI043103; AI790593; carbonyl reductase; CR; D14Ucla2; DCR-AKL; dehydrogenase/reductase (SDR family) member 4; dehydrogenase/reductase (SDR family) member 4 like 2A; dehydrogenase/reductase 4; dehydrogenase/reductase SDR family member 4; DHRS4; mouNRDR; NADH-dependent retinal reductase; NADPH-dependent carbonyl reductase/NADP-retinol dehydrogenase; NADPH-dependent retinol dehydrogenase/reductase; NADP-retinol dehydrogenase; NDRD; NDRD protein; NRDR; Peroxisomal short-chain alcohol dehydrogenase; PH PSCD; PHCR; PSCD; rabNRDR; RRD; SCAD-SRL; SDR25C1; SDR25C2; SDR-SRL; Short chain dehydrogenase/reductase family 25C member 2; short chain dehydrogenase/reductase family 25C, member 1; short chain dehydrogenase/reductase family 25C, member 2; short-chain dehydrogenase/reductase family member 4; UNQ851/PRO1800
Gene Symbols DHRS4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-278 of human DHRS4 (NP_0662842).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10901, 266686, 28200
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.