Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHRS4L1 Polyclonal Antibody
DHRS4L1 Polyclonal Antibody
Supplier: Thermo Scientific PA548394
Description
DHRS4L1 Polyclonal Antibody for Western Blot

Specifications
DHRS4L1 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
P0CG22 | |
DHRS4L1 | |
KLH conjugated synthetic peptide between 222-250 amino acids from the C-terminal region of human DHRS4L1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
DHRS4L1 | |
dehydrogenase/reductase (SDR family) member 4 like 1; dehydrogenase/reductase 4 like 1; DHRS4L1; putative dehydrogenase/reductase SDR family member 4-like 1; Putative dehydrogenase/reductase SDR family member 4-like 2; SDR25C4; short chain dehydrogenase/reductase family 25C member 4; short chain dehydrogenase/reductase family 25C, member 4 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
728635 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction