Learn More
Invitrogen™ DHRS4L2 Monoclonal Antibody (OTI8A2)

Description
DHRS4L2 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | DHRS4L2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI8A2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DHRS4L2 |
| Gene Accession No. | Q6PKH6 |
| Gene Alias | dehydrogenase/reductase (SDR family) member 4 like 2; dehydrogenase/reductase (SDR family) member 4 like 2A3; dehydrogenase/reductase 4 like 2; dehydrogenase/reductase SDR family member 4-like 2; DHRS4L2; NADP(H)-dependent retinol dehydrogenase/reductase short isoform-like protein; NADPH-dependent retinol dehydrogenase/reductase-like protein 2; SDR25C3; short chain dehydrogenase/reductase family 25C member 3; short chain dehydrogenase/reductase family 25C, member 3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.