Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHRS7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533469
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533469 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533469 Supplier Invitrogen™ Supplier No. PA533469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Bat, Horse (100%) Marmoset, Elephant, Pig (94%).

Short-chain dehydrogenases/reductases (SDRs), such as DHRS7, catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids (Haeseleer and Palczewski, 2000 [PubMed 10800688]).[supplied by OMIM, Jun 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene DHRS7
Gene Accession No. Q9Y394
Gene Alias 2310016E22Rik; 5730564L20Rik; AW061210; CGI-86; dehydrogenase/reductase (SDR family) member 7; dehydrogenase/reductase 7; dehydrogenase/reductase SDR family member 7; Dhrs7; DHRS7A; Retdsr4; retinal short-chain dehydrogenase/reductase 4; Retsdr4; RGD1565002; SDR34C1; short chain dehydrogenase/reductase family 34C member 1; short chain dehydrogenase/reductase family 34C, member 1; similar to Dehydrogenase/reductase SDR family member 7 precursor (Retinal short-chain dehydrogenase/reductase 4); similar to Dehydrogenase/reductase SDR family member 7 precursor (Retinal short-chain dehydrogenase/reductase 4) (retSDR4); uncharacterized protein LOC299135; UNQ285/PRO3448
Gene Symbols DHRS7
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from internal region of human DHRS7.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100060062, 51635
Target Species Human, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.