Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHRS7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599104
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599104 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599104 Supplier Invitrogen™ Supplier No. PA599104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DHRS7 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Short-chain dehydrogenases/reductases (SDRs), such as DHRS7, catalyze the oxidation/reduction of a wide range of substrates, including retinoids and steroids (Haeseleer and Palczewski, 2000 [PubMed 10800688]).[supplied by OMIM, Jun 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS7
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DHRS7
Gene Accession No. Q9Y394
Gene Alias 2310016E22Rik; 5730564L20Rik; AW061210; CGI-86; dehydrogenase/reductase (SDR family) member 7; dehydrogenase/reductase 7; dehydrogenase/reductase SDR family member 7; Dhrs7; DHRS7A; Retdsr4; retinal short-chain dehydrogenase/reductase 4; Retsdr4; RGD1565002; SDR34C1; short chain dehydrogenase/reductase family 34C member 1; short chain dehydrogenase/reductase family 34C, member 1; similar to Dehydrogenase/reductase SDR family member 7 precursor (Retinal short-chain dehydrogenase/reductase 4); similar to Dehydrogenase/reductase SDR family member 7 precursor (Retinal short-chain dehydrogenase/reductase 4) (retSDR4); uncharacterized protein LOC299135; UNQ285/PRO3448
Gene Symbols DHRS7
Host Species Rabbit
Immunogen Recombinant Human Dehydrogenase/reductase SDR family member 7 protein (29-339AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51635
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.