Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DHRS7B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552916
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552916 Supplier Invitrogen™ Supplier No. PA552916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATQAFFDCLR AEMEQYEIEV TVISPGYIHT NLSVNAITAD GSRYGVMDTT TAQGRSPVEV AQDVLAAVGK KKKDVILADL LPSLAVYLRT LAPGLFFSLM ASRARKERKS KNS Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

Putative oxidoreductase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DHRS7B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DHRS7B
Gene Accession No. Q6IAN0
Gene Alias BC003479; C79874; C80074; CGI-93; dehydrogenase/reductase (SDR family) member 7B; dehydrogenase/reductase 7B; dehydrogenase/reductase SDR family member 7B; Dhrs7b; PexRAP; RGD1311243; SDR family dehydrogenase/reductase member 7B; SDR32C1; short chain dehydrogenase/reductase family 32C, member 1; short-chain dehydrogenase/reductase family 32C member 1; UNQ212/PRO238
Gene Symbols DHRS7B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DHRS7B. Recombinant protein control fragment (Product #RP-90086).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25979
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.