Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHRS7B Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548554
Description
Putative oxidoreductase.
Specifications
DHRS7B | |
Polyclonal | |
Unconjugated | |
DHRS7B | |
Dehydrogenase/reductase SDR family member 7B, 11--, DHRS7B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25979 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.46 mg/mL | |
PBS with 0.09% sodium azide | |
Q6IAN0 | |
DHRS7B | |
KLH conjugated synthetic peptide between 133-161 amino acids from the Central region of human DHRS7B. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction